Learn More
Sino Biological Influenza A H1N2 (A/Swine/Spain/SF12091/2007) Neuraminidase / NA (His Tag)
Supplier:  Sino Biological 40746V07H100
Encompass_Preferred
A DNA sequence encoding the influenza A virus (A/Swine/Spain/SF12091/2007 (H1N2)) neuraminidase (CCQ71972) (His36-Ile469), termed as NA, was fused with a N-terminal polyhistidine tag.
Catalog No. 50-255-3602