Learn More
Sino Biological Influenza A H1N2 (A/swine/Alberta/SD0217/2017) Neuraminidase / NA (His Tag)
Supplier:  Sino Biological 40750V07H100
Encompass_Preferred
A DNA sequence encoding the influenza A virus (A/swine/Alberta/SD0217/2017 (H1N2)) neuraminidase (QAX25989) (His36-Ile469), termed as NA, was fused with a N-terminal polyhistidine tag.
Catalog No. 50-255-3598