Learn More
Sino Biological Influenza A H1N3 Hemagglutinin / HA Antibody, Mouse MAb
Supplier:  Sino Biological 11685MM0720
Encompass_Preferred
This antibody was produced from a hybridoma resulting from the fusion of a mouse myeloma with B cells obtained from a mouse immunized with purified, recombinant Influenza A virus H1N3 Hemagglutinin. The IgG fraction of the cell culture supernatant was purified by Protein A affinity chromatography.
Catalog No. 50-241-1842