Learn More
Sino Biological Influenza A H1N3 Hemagglutinin / HA Antibody, Rabbit PAb, Antigen Affinity Purified
Supplier:  Sino Biological 11685RP02100
Encompass_Preferred
Produced in rabbits immunized with purified, recombinant Influenza A virus H1N3 Hemagglutinin (rV Influenza A virus H1N3 Hemagglutinin; Catalog#11685-V08H; Met 1-Gln 529; ABB20429.1). Influenza A virus H1N3 Hemagglutinin specific IgG was purified by Influenza A virus H1N3 Hemagglutinin affinity chromatography.
Catalog No. 50-241-1841