Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC H1PVAT | 351438-49-8 | 96.4% | 417.42 g·mol⁻¹ | C19H14F3N5OS | 5MG
SDP

Catalog No. 5000238995
Encompass_Preferred

H1PVAT is a small-molecule antiviral research compound that potently and selectively inhibits poliovirus serotypes PV-1, PV-2, and PV-3 by interfering with early stages of viral replication. It is supplied for laboratory research use and is commonly used to study enterovirus capsid interactions and antiviral mechanisms.

  • Potent inhibitor of poliovirus serotypes PV-1, PV-2, and PV-3.
  • Inhibits early stage of viral replication.
  • Suitable for in vitro antiviral and enterovirus research.
  • Molecular formula C19H14F3N5OS.
  • Molecular weight 417.42 g·mol⁻¹.
  • Reported purity approximately 96.4%.

Catalog No. 50-002-38995 Supplier Medchemexpress LLC Supplier No. HY123495MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

H1PVAT is a small-molecule antiviral research compound that potently and selectively inhibits poliovirus serotypes PV-1, PV-2, and PV-3 by interfering with early stages of viral replication. It is supplied for laboratory research use and is commonly used to study enterovirus capsid interactions and antiviral mechanisms.

  • Potent inhibitor of poliovirus serotypes PV-1, PV-2, and PV-3.
  • Inhibits early stage of viral replication.
  • Suitable for in vitro antiviral and enterovirus research.
  • Molecular formula C19H14F3N5OS.
  • Molecular weight 417.42 g·mol⁻¹.
  • Reported purity approximately 96.4%.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.