Learn More
Medchemexpress LLC H2-003 | 1060438-30-3 | 99.5% | 446.50 | 5 MG
SDP
Catalog No. 5000400535
Encompass_Preferred
H2-003 is a selective human DGAT2 inhibitor that inhibits triglyceride (TG) biosynthesis. It also effectively inhibits lipid droplet formation in 3T3-L1 cells. H2-003 can be used for research on DGAT2 and TG-related metabolic diseases.
  • Selective human DGAT2 inhibitor
  • Inhibits triglyceride (TG) biosynthesis
  • Effectively inhibits lipid droplet formation in 3T3-L1 cells
  • Used for research on DGAT2 and TG-related metabolic diseases
Catalog No. 50-004-00535 Supplier Medchemexpress LLC Supplier No. HY231995MG
H2-003 is a selective human DGAT2 inhibitor that inhibits triglyceride (TG) biosynthesis. It also effectively inhibits lipid droplet formation in 3T3-L1 cells. H2-003 can be used for research on DGAT2 and TG-related metabolic diseases.
  • Selective human DGAT2 inhibitor
  • Inhibits triglyceride (TG) biosynthesis
  • Effectively inhibits lipid droplet formation in 3T3-L1 cells
  • Used for research on DGAT2 and TG-related metabolic diseases