Learn More
ABclonal Technology H2-Ab1 Rabbit mAb
SDP
Catalog No. 5000010250
Encompass_Preferred
Enables several functions including peptide antigen binding activity protein antigen binding activity and toxic substance binding activity Involved in several processes including B cell affinity maturation cellular response to interferon-gamma and positive regulation of T-helper 1 type immune response Acts upstream of or within antigen processing and presentation of exogenous peptide antigen via MHC class II and immune response Located in Golgi apparatus endosome and external side of plasma membrane Is integral component of membrane Part of MHC class II protein complex Is expressed in lung metanephros and thymus primordium Orthologous to human HLA-DQB2 (major histocompatibility complex class II DQ beta 2)
Catalog No. 50-000-10250 Supplier ABclonal Technology Supplier No. A2465020UL
Enables several functions including peptide antigen binding activity protein antigen binding activity and toxic substance binding activity Involved in several processes including B cell affinity maturation cellular response to interferon-gamma and positive regulation of T-helper 1 type immune response Acts upstream of or within antigen processing and presentation of exogenous peptide antigen via MHC class II and immune response Located in Golgi apparatus endosome and external side of plasma membrane Is integral component of membrane Part of MHC class II protein complex Is expressed in lung metanephros and thymus primordium Orthologous to human HLA-DQB2 (major histocompatibility complex class II DQ beta 2)