Learn More
Alkali Scientific H2505-40 SureTemp 40L Lab Incu
SDP
Catalog No. 503029599
Encompass_Preferred
Precision temperature control up to 75C Dual-Convection technology excellent uniformity and recovery Includes SureChecktemperature logger and software Stainless steel chamber with internal power outlet The new H2505-40 SureTemp lab incubator from Alkali Scientificis simple to use for general lab incubation but also equipped with the SureCheckdata logging system for verification of chamber temperature when a high level of precision is needed The new SureTempDual Convection Incubator series provides the highest level of temperature control with uniformity and stability levels that are unmatched by competitive models in the same class Each incubator is equipped with the SureCheckdata logging system for verification of chamber temperature The SureCheck system records temperature data points as often as every 60 seconds and stores this information on a flash drive The data can then be viewed on any computer capable of reading a csv file from the flash drive The recorded data can also
Catalog No. 50-302-9599 Supplier Alkali Scientific Supplier No. RS250540
Precision temperature control up to 75C Dual-Convection technology excellent uniformity and recovery Includes SureChecktemperature logger and software Stainless steel chamber with internal power outlet The new H2505-40 SureTemp lab incubator from Alkali Scientificis simple to use for general lab incubation but also equipped with the SureCheckdata logging system for verification of chamber temperature when a high level of precision is needed The new SureTempDual Convection Incubator series provides the highest level of temperature control with uniformity and stability levels that are unmatched by competitive models in the same class Each incubator is equipped with the SureCheckdata logging system for verification of chamber temperature The SureCheck system records temperature data points as often as every 60 seconds and stores this information on a flash drive The data can then be viewed on any computer capable of reading a csv file from the flash drive The recorded data can also