Learn More
Alkali Scientific H2505-70 SureTemp 70L Lab Incu
SDP
Catalog No. 503029615
Encompass_Preferred
The new Alkali ScientificH2505-70 SureTemp lab incubator from Alkali Scientificis simple to use for general lab incubation but also equipped with the SureCheckdata logging system for verification of chamber temperature when a high level of precision is needed The SureCheck system records temperature data points as often as every 60 seconds and stores this information on a flash drive The data can then be viewed and saved using the included the SureCheck software The recorded data can also be used to plot temperature points on a graph or exported to a spread sheet program for verification or storage/record keeping The Alkali ScientificH2505-70 SureTemp incubator offers Dual-Convection technology allowing the user to choose between mechanical convection mode for exceptional temperature uniformity or gravity convection mode for longer incubations where airborne contamination or drying of samples may be an issue The heating elements are strategically located on multiple sides of the c
Catalog No. 50-302-9615 Supplier Alkali Scientific Supplier No. RS250570
The new Alkali ScientificH2505-70 SureTemp lab incubator from Alkali Scientificis simple to use for general lab incubation but also equipped with the SureCheckdata logging system for verification of chamber temperature when a high level of precision is needed The SureCheck system records temperature data points as often as every 60 seconds and stores this information on a flash drive The data can then be viewed and saved using the included the SureCheck software The recorded data can also be used to plot temperature points on a graph or exported to a spread sheet program for verification or storage/record keeping The Alkali ScientificH2505-70 SureTemp incubator offers Dual-Convection technology allowing the user to choose between mechanical convection mode for exceptional temperature uniformity or gravity convection mode for longer incubations where airborne contamination or drying of samples may be an issue The heating elements are strategically located on multiple sides of the c