Learn More
Aviva Systems Biology Corp H2AFX Antibody - N-terminal region (ARP58280_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP58280_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against H2AFX. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the N terminal region of human H2AFX. Alternate names: H2A.X, H2A/X, H2AXApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rat, Zebrafish
Catalog No. 50-014-62273