Learn More
Aviva Systems Biology Corp H2AFY Antibody - middle region (ARP58283_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP58283_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against H2AFY. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the middle region of human H2AFY. Alternate names: H2A.y, H2A/y, H2AF12M, H2AFJ, MACROH2A1.1, mH2A1, macroH2A1.2ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Pig, Rabbit, Rat
Catalog No. 50-014-62276