Learn More
Aviva Systems Biology Corp H2AFZ Antibody - C-terminal region (ARP61353_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP61353_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against H2AFZ. It was validated on Western Blot. . Alternate names: H2A.z, H2A/z, H2AZ, MGC117173ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat, Sheep, Zebrafish
Catalog No. 50-287-349