Learn More
StatLab H2Blue Absorbant Beads
SDP
Catalog No. 22000037
Change view
Click to view available options
Form:
Pellet
Catalog No. Form
22-000-037 Pellet
1 options
Harmful if swallowed, in contact with skin, or if inhaled
Catalog No. 22-000-037 Supplier StatLab Supplier No. FIR0185
Add to Cart
Add to Cart
Harmful if swallowed, in contact with skin, or if inhaled
Instead of discarding your clearing agent so often, double the volume of water it can retain and extend its useful life with H2Blue
  • Just add 10 g of H2Blue to 500 mL of Xylene or Clearify, and you can nearly double it's usage between changes
  • H2Blue contains blue indicator beads that lose their color when the maximum amount of water is absorbed
  • If water is shortening the life your clearing agent, clear it up with H2Blue!
Specifications
Product Type Clearing Agent
Form Pellet