Learn More
Selleck Chemical LLC H2DCFDA S9687-100MG
SDP
Catalog No. 5000024904
Encompass_Preferred
H2DCFDA (DCFH-DA) is a cell-permeable probe used to detect intracellular reactive oxygen species (ROS) production It is de-esterified intracellularly and turns to highly fluorescent 2 7 -dichlorofluorescein upon oxidation It is used in sensitive and rapid quantitation of oxygen-reactive species in response to oxidative metabolism microplate assay for detecting oxidative products in phagocytic cells and quantitative multiwell myeloid differentiation assay (Ex/Em 488/525 nm)
Catalog No. 50-000-24904 Supplier Selleck Chemical LLC Supplier No. S9687100MG
H2DCFDA (DCFH-DA) is a cell-permeable probe used to detect intracellular reactive oxygen species (ROS) production It is de-esterified intracellularly and turns to highly fluorescent 2 7 -dichlorofluorescein upon oxidation It is used in sensitive and rapid quantitation of oxygen-reactive species in response to oxidative metabolism microplate assay for detecting oxidative products in phagocytic cells and quantitative multiwell myeloid differentiation assay (Ex/Em 488/525 nm)