Learn More
Novus Biologicals™ H2DCFDA Inhibitor

Description
Biological Activity: Fluorescent ROS indicator; becomes fluorescent when oxidized. Excitation wavelength: 490nm, emission: 520nm. Cell permeable.
Storage: Store at 4°C.
Note: Inhibitors are guranteed for 1 year from date of receipt.
Specifications
Specifications
| Description | CAS: 4091-99-0 |
| Formulation | C24H16Cl2O7 |
| For Use With (Application) | Used to indicate ROS |
| Quantity | 100 mg |
| Product Type | Staining solution |
| Molecular Weight (g/mol) | 487.29g/mol |
| Solubility | Soluble to 100mM in DMSO, 10mM in ethanol |
| Content And Storage | -20°C |
| Color | White |
For Research Use Only
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.