Learn More
Medchemexpress LLC (H2IPr)(Cl)2Ru=CH(o-iPrOC6H4) | 635679-24-2 | 710.78 | 250 MG
SDP
Catalog No. 5000366172
Encompass_Preferred
(H2IPr)(Cl)2Ru=CH(o-iPrOC6H4) is a highly efficient catalyst designed to promote the metathesis reaction of olefins. This compound demonstrates excellent selectivity in transition metal catalysis, making it suitable for synthesizing biologically active compounds and showing potential application value in compound development.
  • Highly efficient catalyst
  • Effectively promotes olefin metathesis reactions
  • Exhibits excellent selectivity in transition metal catalysis
  • Suitable for the synthesis of biologically active compounds
  • Potential application value in compound development
Catalog No. 50-003-66172 Supplier Medchemexpress LLC Supplier No. 250MGHYW854275
(H2IPr)(Cl)2Ru=CH(o-iPrOC6H4) is a highly efficient catalyst designed to promote the metathesis reaction of olefins. This compound demonstrates excellent selectivity in transition metal catalysis, making it suitable for synthesizing biologically active compounds and showing potential application value in compound development.
  • Highly efficient catalyst
  • Effectively promotes olefin metathesis reactions
  • Exhibits excellent selectivity in transition metal catalysis
  • Suitable for the synthesis of biologically active compounds
  • Potential application value in compound development