Learn More
Enzo Life Sciences Hydrogen peroxide chemiluminescent detection kit (1x96wells)
SDP
Catalog No. 50658604
Encompass_Preferred
The Hydrogen Peroxide chemiluminescent detection kit provides a fast and higher throughput 96-well plate based homogeneous assay for the measurement of hydrogen peroxide. This kit offers more sensitivity over the Hydrogen Peroxide colorimetric detection kit (Cat. #ADI-907-015) but is not recommended for samples that contain phenol red or hemoglobin. Sensitivity: 2.39 ng/ml (range 2.44 - 2,500 ng/ml). Applications: Chemiluminescence. Species Reactivity: Species independent. Contents: White Microtiter plate, Standard, Substrate, Trigger solution
Catalog No. 50-658-604 Supplier Enzo Life Sciences Supplier No. ADI907012
The Hydrogen Peroxide chemiluminescent detection kit provides a fast and higher throughput 96-well plate based homogeneous assay for the measurement of hydrogen peroxide. This kit offers more sensitivity over the Hydrogen Peroxide colorimetric detection kit (Cat. #ADI-907-015) but is not recommended for samples that contain phenol red or hemoglobin. Sensitivity: 2.39 ng/ml (range 2.44 - 2,500 ng/ml). Applications: Chemiluminescence. Species Reactivity: Species independent. Contents: White Microtiter plate, Standard, Substrate, Trigger solution