Learn More
Apexbio Technology LLC H2O2 COL KIT LIPID-250mg
SDP
Catalog No. 5000567515
Encompass_Preferred
Hydrogen Peroxide Colorimetric Assay Kit (Lipid-Compatible) is utilized in biomedical research to measure hydrogen peroxide levels in lipid-rich samples Originating from advancements in assay technology it targets oxidative stress pathways by detecting hydrogen peroxide a key reactive oxygen species The kit employs a colorimetric mechanism where hydrogen peroxide reacts with a specific reagent resulting in a color change measurable at a defined wavelength Common in vitro activity is observed in the nanomolar to low micromolar range facilitating high sensitivity and specificity It is widely used in cell models and animal studies to investigate oxidative stress lipid peroxidation and in drug screening to evaluate antioxidant efficacy Experimental concentrations typically depend on the specific research design ensuring adaptability across diverse experimental contexts
Catalog No. 50-005-67515 Supplier Apexbio Technology LLC Supplier No. K2310250
Hydrogen Peroxide Colorimetric Assay Kit (Lipid-Compatible) is utilized in biomedical research to measure hydrogen peroxide levels in lipid-rich samples Originating from advancements in assay technology it targets oxidative stress pathways by detecting hydrogen peroxide a key reactive oxygen species The kit employs a colorimetric mechanism where hydrogen peroxide reacts with a specific reagent resulting in a color change measurable at a defined wavelength Common in vitro activity is observed in the nanomolar to low micromolar range facilitating high sensitivity and specificity It is widely used in cell models and animal studies to investigate oxidative stress lipid peroxidation and in drug screening to evaluate antioxidant efficacy Experimental concentrations typically depend on the specific research design ensuring adaptability across diverse experimental contexts