Learn More
Apexbio Technology LLC H2O2 COL KIT WATER-250mg
SDP
Catalog No. 5000567666
Encompass_Preferred
Hydrogen Peroxide Colorimetric Assay Kit (Water-Compatible) is utilized in biomedical research for detecting and quantifying hydrogen peroxide in various samples originating from oxidative stress studies The mechanism involves a colorimetric reaction that produces a quantifiable color change when hydrogen peroxide is present typically in nanomolar to low micromolar concentrations This kit is widely used for in vitro applications including cell models animal studies and drug screening allowing researchers to assess oxidative damage or antioxidant capabilities Experimental concentrations typically depend on the specific research design
Catalog No. 50-005-67666 Supplier Apexbio Technology LLC Supplier No. K2309250
Hydrogen Peroxide Colorimetric Assay Kit (Water-Compatible) is utilized in biomedical research for detecting and quantifying hydrogen peroxide in various samples originating from oxidative stress studies The mechanism involves a colorimetric reaction that produces a quantifiable color change when hydrogen peroxide is present typically in nanomolar to low micromolar concentrations This kit is widely used for in vitro applications including cell models animal studies and drug screening allowing researchers to assess oxidative damage or antioxidant capabilities Experimental concentrations typically depend on the specific research design