Learn More
Enzo Life Sciences Hydrogen peroxide colorimetric detection kit (1x96wells)
SDP
Supplier:  Enzo Life Sciences ADI907015
Encompass_Preferred
The Hydrogen Peroxide colorimetric detection kit provides a fast and higher throughput 96-well plate based homogeneous assay for the measurement of hydrogen peroxide in a wide variety of samples. Sensitivity: 51.25 ng/ml (range 106.25 - 3,400 ng/ml). Applications: Colorimetric detection, HTS. Species Reactivity: Species independent. Contents: Half-Area Microtiter plate, Standard, Color reagent
Catalog No. 50-658-607