Learn More
Caron Products And Services H202 Sterilization Chamber Prep
Catalog No. NC2684433
Encompass_Preferred
H2O2 sterilization prep, factory installed. For use in 7400/7401/7402/7403 Series incubators. Includes H2O2 sterilization module connections, upgraded system wiring, and control software & hardware. STER301 H2O2 sterilization module sold separately.
Catalog No. NC2684433 Supplier Caron Products And Services Supplier No. STER305
H2O2 sterilization prep, factory installed. For use in 7400/7401/7402/7403 Series incubators. Includes H2O2 sterilization module connections, upgraded system wiring, and control software & hardware. STER301 H2O2 sterilization module sold separately.