Learn More
Cayman Chemical H2S Donr 5a 10mg
SDP
Catalog No. 502952046
Encompass_Preferred
A stable, cysteine-activated H2S donor
Catalog No. 50-295-2046 Supplier Cayman Chemical Supplier No. 1123810 MG
A stable, cysteine-activated H2S donor