Learn More
GENERAL MONITORS H2S SENSOR 0-50PPM
Catalog No. NC9199575
Encompass
H2S MOS Aluminum sensor, 0 - 50 ppm General Monitorsʼ hydrogen sulfide (H2S) sensors are solid state devices, designed and manufactured for long life and fast response. They are selective to H2S and remain unaffected by high concentrations of other substances like hydrogen, sulfur dioxide, and gasoline vapors, which are often present in facilities that process sour gas and crude oil. A high tolerance to a broad range of temperatures and humidity enable these sensors to operate in rugged environments, as does their capacity to withstand exposure to high H2S concentrations over short periods. *Solid-state operation functions in the harshest environments *Highly selective -- does not respond to hydrocarbons -- for low risk of false alarms *Unaffected by over-range exposure reduces need for sensor replacement *Robust mechanical design *Resistant to high humidity and wide range of ambient temperatures *Specific sensors meet ISA-92.0.01 performance standard
Catalog No. NC9199575 Supplier GENERAL MONITORS Supplier No. 504455
H2S MOS Aluminum sensor, 0 - 50 ppm General Monitorsʼ hydrogen sulfide (H2S) sensors are solid state devices, designed and manufactured for long life and fast response. They are selective to H2S and remain unaffected by high concentrations of other substances like hydrogen, sulfur dioxide, and gasoline vapors, which are often present in facilities that process sour gas and crude oil. A high tolerance to a broad range of temperatures and humidity enable these sensors to operate in rugged environments, as does their capacity to withstand exposure to high H2S concentrations over short periods. *Solid-state operation functions in the harshest environments *Highly selective -- does not respond to hydrocarbons -- for low risk of false alarms *Unaffected by over-range exposure reduces need for sensor replacement *Robust mechanical design *Resistant to high humidity and wide range of ambient temperatures *Specific sensors meet ISA-92.0.01 performance standard