Learn More
AnaSpec Histone H3 (1-21), N-Terminal, 1mg
Supplier:  AnaSpec AS61701
Encompass
Histone H3 (1-21), N-Terminal[ARTKQTARKSTGGKAPRKQLA] ; MW 2254.6; (1 mg) This peptide is derived from Histone H3 1-21 amino acids, and is usually used as a substrate for methylation and acetylation assays.
Catalog No. NC0411398