Learn More
Alkali Scientific H3020 Incu-Shaker 20L Stackabl
SDP
Catalog No. 503029516
Encompass_Preferred
Stainless steel chamber provides premier temperature uniformity Proven reliable maintenance-free Incu-Shakermotor drive system Bright intuitive touch-screen controls Instantly change flask clamps with MAGic Clampaccessories Included pull-out adjustable shelf for static incubations The new Alkali Incu-Shaker20L (H3020) introduce a high-throughput space saving shaking incubator system to the Incu-Shaker family Now with stacking capability of up to 2 units the Incu-Shaker20 series are highly space-efficient without compromising on reliability or temperature performance Thoroughly tested for years of reliable performance the same proven brushless induction motor drive is featured as in other Incu-Shaker instruments The high grade stainless-steel internal chambers provide temperature accuracy within 0 1C and industry-leading uniformity within the chamber of 0 5C The Incu-Shaker20L features temperature control from 5C above ambient temperatures up to 60C The 20LR features a built-in pow
Catalog No. 50-302-9516 Supplier Alkali Scientific Supplier No. RSH3020
Add to Cart
Add to Cart
Request a Quote
Stainless steel chamber provides premier temperature uniformity Proven reliable maintenance-free Incu-Shakermotor drive system Bright intuitive touch-screen controls Instantly change flask clamps with MAGic Clampaccessories Included pull-out adjustable shelf for static incubations The new Alkali Incu-Shaker20L (H3020) introduce a high-throughput space saving shaking incubator system to the Incu-Shaker family Now with stacking capability of up to 2 units the Incu-Shaker20 series are highly space-efficient without compromising on reliability or temperature performance Thoroughly tested for years of reliable performance the same proven brushless induction motor drive is featured as in other Incu-Shaker instruments The high grade stainless-steel internal chambers provide temperature accuracy within 0 1C and industry-leading uniformity within the chamber of 0 5C The Incu-Shaker20L features temperature control from 5C above ambient temperatures up to 60C The 20LR features a built-in pow