Learn More
Alkali Scientific H3565-45 45L CO2 Incubator Wit
SDP
Catalog No. 503029679
Encompass_Preferred
Internal microscope option Precision temperature control Excellent accuracy and recovery Heated door and door frame Cell cultures depend on optimized growth conditions in order to thrive With precise temperature and CO control Alkali Scientifics H3565-45 SureTherm45L CO2 Incubator provides the perfect environment for your cultures The Alkali ScientificH3565-45 SureTherm45L CO2 Incubator features a proprietary heat distribution system that combines six sided heating with a low speed internal fan Air is circulated uniformly throughout the chamber without air exchange from the external environment This results in a controlled protected environment with minimal temperature fluctuations (within 0 1C) These are the first and only incubators with our OPTIONAL (patent pending) IncuViewLCI (Live Cell Imaging) technology for viewing cells in real time An internal microscope and large external display allow cells to be viewed without removal from the incubator eliminating the inherent tem
Catalog No. 50-302-9679 Supplier Alkali Scientific Supplier No. RS356545
Add to Cart
Add to Cart
Request a Quote
Internal microscope option Precision temperature control Excellent accuracy and recovery Heated door and door frame Cell cultures depend on optimized growth conditions in order to thrive With precise temperature and CO control Alkali Scientifics H3565-45 SureTherm45L CO2 Incubator provides the perfect environment for your cultures The Alkali ScientificH3565-45 SureTherm45L CO2 Incubator features a proprietary heat distribution system that combines six sided heating with a low speed internal fan Air is circulated uniformly throughout the chamber without air exchange from the external environment This results in a controlled protected environment with minimal temperature fluctuations (within 0 1C) These are the first and only incubators with our OPTIONAL (patent pending) IncuViewLCI (Live Cell Imaging) technology for viewing cells in real time An internal microscope and large external display allow cells to be viewed without removal from the incubator eliminating the inherent tem