Learn More
Alkali Scientific H3710-H Digital Hotplate 10 x
SDP
Catalog No. 503029547
Encompass_Preferred
Large ceramic work surface 10 x 10 inch Safety LED indicates hot surface 5L stir capacity Control actual sample temperature (with optional probe) The Alkali ScientificH3710-H Hotplate provides reliable performance and safe operation for everyday laboratory use Our space efficient 7 x 7-inch hotplate/stirrer series features a durable chemical resistant white ceramic top plate and advanced microprocessor controls The constant monitoring system provides precise control of temperature with rapid heating and faster boiling times In addition the powerful motor drive system ensures efficient mixing in larger and more viscous samples Temperature and mixing speed can be viewed on the large digital display and a safety LED illuminates when the top plate temperature exceeds 50C An external temperature probe can also be used for applications that require precise control of sample temperature The probe provides direct feedback to the microprocessor with constant monitoring and adjustments for
Catalog No. 50-302-9547 Supplier Alkali Scientific Supplier No. RS3710H
Large ceramic work surface 10 x 10 inch Safety LED indicates hot surface 5L stir capacity Control actual sample temperature (with optional probe) The Alkali ScientificH3710-H Hotplate provides reliable performance and safe operation for everyday laboratory use Our space efficient 7 x 7-inch hotplate/stirrer series features a durable chemical resistant white ceramic top plate and advanced microprocessor controls The constant monitoring system provides precise control of temperature with rapid heating and faster boiling times In addition the powerful motor drive system ensures efficient mixing in larger and more viscous samples Temperature and mixing speed can be viewed on the large digital display and a safety LED illuminates when the top plate temperature exceeds 50C An external temperature probe can also be used for applications that require precise control of sample temperature The probe provides direct feedback to the microprocessor with constant monitoring and adjustments for