Learn More
Alkali Scientific H3710-S Digital Magnetic Stirr
SDP
Catalog No. 503029565
Encompass_Preferred
Large 10 x 10-inch white ceramic top plate Maximum stirring capacity 15 L Digital speed and temperature control Safety LED indicates hot surface Control actual sample temperature (with optional probe) Alkali hotplates stirrers and hotplate-stirrers feature an exceptionally durable chemical resistant white ceramic work surface The space-efficient design (7 5 X 11 5 in footprint) makes them ideal for use on crowded benchtops biological safety cabinets and in bio-hoods Advanced microprocessor controls allow quick precise adjustment of speed and temperature The display provides constant visualization of the speed and temperature while a safety LED indicates when the top plate is above 50C In addition an optional probe is available for applications that require precise temperature control When connected the probe provides direct feedback to the microprocessor which continuously adjusts and maintains the selected temperature in the sample within /- 1C With a square 7 in work surface
Catalog No. 50-302-9565 Supplier Alkali Scientific Supplier No. RS3710S
Large 10 x 10-inch white ceramic top plate Maximum stirring capacity 15 L Digital speed and temperature control Safety LED indicates hot surface Control actual sample temperature (with optional probe) Alkali hotplates stirrers and hotplate-stirrers feature an exceptionally durable chemical resistant white ceramic work surface The space-efficient design (7 5 X 11 5 in footprint) makes them ideal for use on crowded benchtops biological safety cabinets and in bio-hoods Advanced microprocessor controls allow quick precise adjustment of speed and temperature The display provides constant visualization of the speed and temperature while a safety LED indicates when the top plate is above 50C In addition an optional probe is available for applications that require precise temperature control When connected the probe provides direct feedback to the microprocessor which continuously adjusts and maintains the selected temperature in the sample within /- 1C With a square 7 in work surface