Learn More
Medchemexpress LLC HY-136128 5mg Medchemexpress, H3B-120 CAS:2194903-42-7 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY1361285MG
Encompass_Preferred
Medchemexpress, HY-136128 5mg H3B-120 CAS:2194903-42-7 H3B-120 is a highly selective, competitive and allosteric carbamoyl phosphate synthetase 1 (CPS1) inhibitor with an IC50 of 1.5 μM and a Ki of 1.4 μM. H3B-120 has anti-cancer activity. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-203-0397