Learn More
Medchemexpress LLC HY-112611 10mg Medchemexpress, H3B-5942 CAS:2052128-15-9 Purity:>98%
SDP
Catalog No. 501969114
Encompass_Preferred
Medchemexpress, HY-112611 10mg H3B-5942 CAS:2052128-15-9 H3B-5942 is a selective, irreversible and orally active estrogen receptor covalent antagonist, inactivates both wild-type and mutant ERα by targeting Cys530, with Kis of 1 nM and 0.41 nM, respectively. H3B-5942 reduces ERα target gene GREB1, shows potent antitumor activity both in multiple cell lines or animals bearing ERαWT or ERα mutations. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-196-9114 Supplier Medchemexpress LLC Supplier No. HY11261110MG
Medchemexpress, HY-112611 10mg H3B-5942 CAS:2052128-15-9 H3B-5942 is a selective, irreversible and orally active estrogen receptor covalent antagonist, inactivates both wild-type and mutant ERα by targeting Cys530, with Kis of 1 nM and 0.41 nM, respectively. H3B-5942 reduces ERα target gene GREB1, shows potent antitumor activity both in multiple cell lines or animals bearing ERαWT or ERα mutations. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.