Learn More
Selleck Chemical LLC H3B-6527 2mg 1702259-66-2
SDP
Supplier:  Selleck Chemical LLC S86752MG
Encompass_Preferred
H3B-6527 is a highly selective covalent FGFR4 inhibitor with an IC50 value of <1.2 nM and at least 250-fold selectivity over FGFR1-3 (IC50 values of 320, 1,290 and 1,060 nM respectively). *For Research & Development use only.  Product is not intended for drug, household, or human consumption.
Catalog No. 50-194-8146