Learn More
TARGETMOL CHEMICALS INC H3B-6527 5MG
Supplier: TARGETMOL CHEMICALS INC T77385MG
Also available in 1 mg 2 mg 10 mg 25 mg 50 mg 100 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. H3B-6527 Is a Potent and Selective Inhibitor of FGFR4 in FGF19-Driven Hepatocellular Carcinoma with antineoplastic activity. purity: 99%