Learn More
Medchemexpress LLC H3B-6545 | 2052130-80-8 | 99.4% | 567.58 | 10 MG
SDP
Catalog No. 5000436658
Encompass_Preferred
H3B-6545 is an oral, selective estrogen receptor covalent antagonist (SERCA) for the research of metastatic ER-positive, HER2-negative breast cancer. It is a first-in-class selective ER covalent antagonist (SERCA) that inactivates both wild-type and mutant ERα by targeting C530 and enforcing a unique antagonist conformation.
  • Inactivates wild-type and mutant ERα by targeting C530.
  • Enforces a unique antagonist conformation.
  • Used for research of metastatic ER-positive, HER2-negative breast cancer.
  • Shows potent activity in MCF-7 xenograft model.
  • Well tolerated in rats and monkeys across a broad dose range.
Catalog No. 50-004-36658 Supplier Medchemexpress LLC Supplier No. HY11259610MG
H3B-6545 is an oral, selective estrogen receptor covalent antagonist (SERCA) for the research of metastatic ER-positive, HER2-negative breast cancer. It is a first-in-class selective ER covalent antagonist (SERCA) that inactivates both wild-type and mutant ERα by targeting C530 and enforcing a unique antagonist conformation.
  • Inactivates wild-type and mutant ERα by targeting C530.
  • Enforces a unique antagonist conformation.
  • Used for research of metastatic ER-positive, HER2-negative breast cancer.
  • Shows potent activity in MCF-7 xenograft model.
  • Well tolerated in rats and monkeys across a broad dose range.