Learn More
Medchemexpress LLC H3B-6545 | 2052130-80-8 | 99.4% | 567.58 | 50 MG
SDP
Catalog No. 5000437397
Encompass_Preferred
H3B-6545 is an oral, selective estrogen receptor covalent antagonist (SERCA) for the research of metastatic ER-positive, HER2-negative breast cancer. It is for research use only and not sold to patients.
  • Oral, selective estrogen receptor covalent antagonist (SERCA).
  • Designed for research into metastatic ER-positive, HER2-negative breast cancer.
  • Potently antagonizes wild-type and mutant ERα in biochemical and cell-based assays.
  • Inactivates wild-type and mutant ERα by targeting C530.
  • Inhibits ERαWT activity and suppresses proliferation of ERαWT-positive breast cancer cell lines.
  • Demonstrates superior efficacy to fulvestrant and tamoxifen in xenograft and patient-derived xenograft models.
  • Well tolerated across a broad dose range in rats and monkeys.
Catalog No. 50-004-37397 Supplier Medchemexpress LLC Supplier No. HY11259650MG
H3B-6545 is an oral, selective estrogen receptor covalent antagonist (SERCA) for the research of metastatic ER-positive, HER2-negative breast cancer. It is for research use only and not sold to patients.
  • Oral, selective estrogen receptor covalent antagonist (SERCA).
  • Designed for research into metastatic ER-positive, HER2-negative breast cancer.
  • Potently antagonizes wild-type and mutant ERα in biochemical and cell-based assays.
  • Inactivates wild-type and mutant ERα by targeting C530.
  • Inhibits ERαWT activity and suppresses proliferation of ERαWT-positive breast cancer cell lines.
  • Demonstrates superior efficacy to fulvestrant and tamoxifen in xenograft and patient-derived xenograft models.
  • Well tolerated across a broad dose range in rats and monkeys.