Learn More
Aviva Systems Biology Corp HIST3H3 Antibody - N-terminal region (AVARP01011_T100) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp AVARP01011_T100
Encompass_Preferred
This is a rabbit polyclonal antibody against H3F3A. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the N terminal region of human HIST3H3. Alternate names: H3t, H3.4, H3/g, H3FT, H3F3AApplicationWB. Species reactivity: Human
Catalog No. 50-014-62464