Learn More
Aviva Systems Biology Corp H3f3a Antibody - N-terminal region (ARP60983_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP60983_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against H3f3a. It was validated on Western Blot. . Alternate names: H3.3A, H3f3b, MGC102150ApplicationWB. Species reactivity: Dog, Pig, Rat, Zebrafish
Catalog No. 50-287-223