Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC H3K4(Me2) (1-20) | 99.9% | 2211.61 | 5 MG
SDP

Catalog No. 5000424428
Encompass_Preferred

H3K4(Me2) (1-20) is a histone peptide that regulates the recovery of protein biosynthesis and homeostasis following DNA damage.

  • Solid appearance
  • White to off-white color
  • Soluble in H2O at 25 mg/mL
  • Recommended storage for powder: -80°C for 2 years or -20°C for 1 year
  • Recommended storage in solvent: -80°C for 6 months or -20°C for 1 month

Catalog No. 50-004-24428 Supplier Medchemexpress LLC Supplier No. HYP22565MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

H3K4(Me2) (1-20) is a histone peptide that regulates the recovery of protein biosynthesis and homeostasis following DNA damage.

  • Solid appearance
  • White to off-white color
  • Soluble in H2O at 25 mg/mL
  • Recommended storage for powder: -80°C for 2 years or -20°C for 1 year
  • Recommended storage in solvent: -80°C for 6 months or -20°C for 1 month

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.