Learn More
EPICYPHER INC H3K4me3 Antibody, SNAP-Certified™ for CUT&RUN and CUT&Tag, 100 µg
SDP
Catalog No. NC2960591
Encompass
This H3K4me3 (histone H3 trimethylated at lysine 4) antibody meets EpiCypher’s lot-specific SNAP-Certified™ criteria for specificity and efficient target enrichment in both CUT&RUN and CUT&Tag applications. This requires <20% cross-reactivity to related histone PTMs determined using the SNAP-CUTANA™ K-MetStat Panel of spike-in controls (EpiCypher 19-1002, Figures 1 and 5). High target efficiency is confirmed by consistent genomic enrichment at varying cell inputs: 500k and 50k cells in CUT&RUN (Figures 2-3); 100k and 10k cells in CUT&Tag (Figures 6-7). High efficiency antibodies display similar peak structures (Figures 3 and 7) and highly conserved genome-wide signal (Figures 2 and 6) even at reduced cell numbers. This antibody targets histone H3K4me3, which is enriched at active promoters near transcription start sites (TSS) and promotes gene activation.
Catalog No. NC2960591 Supplier EPICYPHER INC Supplier No. 130060
This H3K4me3 (histone H3 trimethylated at lysine 4) antibody meets EpiCypher’s lot-specific SNAP-Certified™ criteria for specificity and efficient target enrichment in both CUT&RUN and CUT&Tag applications. This requires <20% cross-reactivity to related histone PTMs determined using the SNAP-CUTANA™ K-MetStat Panel of spike-in controls (EpiCypher 19-1002, Figures 1 and 5). High target efficiency is confirmed by consistent genomic enrichment at varying cell inputs: 500k and 50k cells in CUT&RUN (Figures 2-3); 100k and 10k cells in CUT&Tag (Figures 6-7). High efficiency antibodies display similar peak structures (Figures 3 and 7) and highly conserved genome-wide signal (Figures 2 and 6) even at reduced cell numbers. This antibody targets histone H3K4me3, which is enriched at active promoters near transcription start sites (TSS) and promotes gene activation.