Learn More
Epigentek Inc EpiQuik Histone Methyltransferase Activity/Inhibition Assay Kit (H3K9) (48 assays)
SDP
Supplier:  Epigentek Inc P30031
Encompass_Preferred
This kit is a convenient set of tools that allows the experimenter to measure the activity or inhibition of individual histone methyltransferase that specifically target histone H3 at lysine 9 (H3-K9). The kit is ready-to-use and provides all the essential components needed to carry out a successful HMT activity/inhibition experiment without the need for radioactivity or any special equipment.
Catalog No. NC0006557