Learn More
Sino Biological Influenza A H3N1 (A/swine/England/704563/1995) Hemagglutinin / HA Protein (His Tag)
Supplier:  Sino Biological 40684V08H100
Encompass_Preferred
A DNA sequence encoding the Influenza A virus (A/swine/England/704563/1995 (H3N1)) hemagglutinin (AFR76416) (Met1-Ser525), termed as HA, was expressed with a polyhistidine tag at the C-terminus.
Catalog No. 50-255-3653