Learn More
Creative Biomart Recombinant HBV Pre-S Antigen, His-tagged
Supplier:  Creative Biomart H3N2308
Encompass_Preferred
Recombinant Hepatitis B Virus Pre-S produced in E. coli is a full length Pre-S of HBV subtype adw2, containing 174 amino acids, having a molecular weight of about 25 kDa including a vector sequence an
Catalog No. 50-208-4686