Learn More
Sino Biological Influenza A H3N2 (A/Brisbane/10/2007) Hemagglutinin / HA Antibody, Mouse MAb
Supplier:  Sino Biological 11056MM01100
Encompass_Preferred
This antibody was produced from a hybridoma resulting from the fusion of a mouse myeloma with B cells obtained from a mouse immunized with purified, recombinant Influenza A H3N2 (A/Brisbane/10/2007) Hemagglutinin / HA ( Catalog#11056-V08H; ABW23353.1; Met1-Ile531). The IgG fraction of the cell culture supernatant was purified by Protein A affinity chromatography.
Catalog No. 50-255-4916