Learn More
Sino Biological Influenza A H3N2 (A/Brisbane/10/2007) Hemagglutinin / HA Antibody, Rabbit MAb
Supplier:  Sino Biological 11056R006100
Encompass_Preferred
This antibody was obtained from a rabbit immunized with purified, recombinant Influenza A H3N2 (A/Brisbane/10/2007) Hemagglutinin / HA ( Catalog#11056-V08H; ABW23353.1; Met1-Ile531).
Catalog No. 50-255-4912