Learn More
Sino Biological Influenza A H3N2 Hemagglutinin / HA Antibody, Rabbit PAb, Antigen Affinity Purified
Supplier:  Sino Biological 11056RP02100
Encompass_Preferred
Produced in rabbits immunized with purified, recombinant H3N2 (A/Brisbane/10/2007) HA / Hemagglutinin (Catalog#11056-V08H; ABW23353.1; Met 1-Ile 531). H3N2 (A/Brisbane/10/2007) HA / Hemagglutinin specific IgG was purified by H3N2 (A/Brisbane/10/2007) HA / Hemagglutinin affinity chromatography.
Catalog No. 50-241-1761