Learn More
Sino Biological H3N2 HA PAb 20ul (Sino Biological)
Supplier:  Sino Biological 11056T6220
Encompass_Preferred
Produced in rabbits immunized with purified, recombinant H3N2 (A/Brisbane/10/2007) HA / Hemagglutinin (Catalog#11056-V08H; ABW23353.1; Met 1-Ile 531). Total IgG was purified by Protein A affinity chromatography.
Catalog No. 50-237-2480