Learn More
Sino Biological H3N2 HA PAb 20ul (Sino Biological)
Supplier:  Sino Biological 205076T02100
Encompass_Preferred
Produced in rabbits immunized with purified, recombinant Influenza A H3N2 (A/Aichi/2/1968) Hemagglutinin (HA1 Subunit) ( Catalog#11707-V08H1; AAA43178.1; Met1-Arg345). Total IgG was purified by Protein A affinity chromatography.
Catalog No. 50-237-4157