Learn More
Sino Biological H3N2 HA Protein 100ug (Sino Biological)
Supplier:  Sino Biological 40950V08B100
Encompass_Preferred
A DNA sequence encoding the Influenza A virus ((A/South Australia/34/2019) (H3N2)) hemagglutinin (translated amino acids of EPI1607117) (Met1-Asp529), termed as HA, was expressed with the bacteriophage T4 fibritin and a polyhistidine tag at the C-terminus.
Catalog No. 50-237-2061