Learn More
Sino Biological Influenza A H3N2 (A/swine/Denmark/101501-1/2010) Hemagglutinin / HA Protein (His Tag)
Supplier:  Sino Biological 40688V08H100
Encompass_Preferred
A DNA sequence encoding the Influenza A virus (A/swine/Denmark/101501-1/2010 (H3N2)) hemagglutinin (AIZ95441) (Met1-Leu525), termed as HA, was expressed with a polyhistidine tag at the C-terminus.
Catalog No. 50-255-3650