Learn More
Sino Biological Influenza A H3N2 (A/X-31) Hemagglutinin / HA Protein (ECD, His & AVI Tag)(HPLC-verified)
Supplier:  Sino Biological 40059V49H100
Encompass_Preferred
A DNA sequence encoding the Influenza A virus ((A/X-31) (H3N2)) hemagglutinin (translated amino acids of P03438) (Met1-Trp530was expressed with a polyhistidine tag at the C-terminus.
Catalog No. 50-241-2581