Learn More
Sino Biological Influenza A H3N2 (A/Mexico/InDRE2250/2005) Hemagglutinin / HA Protein (His Tag)
Supplier:  Sino Biological 40703V08H100
Encompass_Preferred
A DNA sequence encoding the Influenza A virus (A/Mexico/InDRE2250/2005 (H3N2)) hemagglutinin (AVI15186) (Met1-Ser525), termed as HA, was expressed with a polyhistidine tag at the C-terminus.
Catalog No. 50-255-3636